Learn More
Electron Microscopy Sciences Paraformaldehyde 4% with Glutaraldehyde 1% in 0.1M Phosphate Buffer, pH 7.0 500 ML
SDP
Catalog No. 501901155
Encompass_Preferred
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. Paraformaldehyde 4 % Glutaraldehyde 1% in 0.1M Phosphate Buffer.
pH: 7.0
Catalog No. 50-190-1155 Supplier Electron Microscopy Sciences Supplier No. 1594980
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. Paraformaldehyde 4 % Glutaraldehyde 1% in 0.1M Phosphate Buffer.
pH: 7.0