Learn More
Aviva Systems Biology Corp PARG Antibody - C-terminal region (ARP63555_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63555_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PARG. It was validated on Western Blot. . Alternate names: FLJ60456, PARG99ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-009