Learn More
Enzo Life Sciences PARG (bovine thymus) (0.1U)
SDP
Supplier:  Enzo Life Sciences ALX202045UC01
Encompass_Preferred
Alternative name: Poly(ADP-ribose) glycohydrolase. Formulation: Liquid. In 50mM K-PO4 buffer, pH 7.5, containing 50mM potassium chloride, 0.1% Triton-x 100, 10% glycerol and 1mM DTT, 10mM β-Mercaptoethanol. Source: Isolated from calf thymus. Long Term Storage: -80°C. Use/Stability: For further dilution use the following enzyme dilution buffer: 50mM KPO4, pH 7.5, 50mM KCl, 10 mM β-mercaptoethanol, 10% glycerol, 1mM DTT, 0.1% Triton. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8048