Learn More
Medchemexpress LLC HY-A0091A 500mg , Pargyline CAS:555-57-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA0091A500MG
Encompass_Preferred
HY-A0091A 500mg Pargyline CAS: 555-57-7 Pargyline is an irreversible monoamine oxidase (MAO) inhibitor with Kis of 13 μM and 0.5 μM for MAO-A and MAO-B, respectively. Pargyline has antihypertensive and anticancer activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1517