Learn More
Cayman Chemical Pargyline (hydrochloride), Purity: ≥98%, CAS: 306-07-0, 1 g
SDP
Catalog No. 502889138
Encompass_Preferred
An irreversible inhibitor of MAO (Kis = 15 and 1.8 μM for MAO-A and MAO-B, respectively); induces a moderate decrease of systolic blood pressure in unanaesthetized hypertensive rats but not normotensive WKR or Sprague-Dawley rats; prevents reactive oxygen species-mediated monocyte hypertrophy at a concentration of 10 µM
Catalog No. 50-288-9138 Supplier Cayman Chemical Supplier No. 100078521 G
An irreversible inhibitor of MAO (Kis = 15 and 1.8 μM for MAO-A and MAO-B, respectively); induces a moderate decrease of systolic blood pressure in unanaesthetized hypertensive rats but not normotensive WKR or Sprague-Dawley rats; prevents reactive oxygen species-mediated monocyte hypertrophy at a concentration of 10 µM