Learn More
Selleck Chemical LLC Pargyline hydrochloride S3690-1g
SDP
Catalog No. 503110750
Encompass_Preferred
Pargyline is an irreversible inhibitor of monoamine oxidase (MAO)-B with Ki values of 13 and 0 5 M for time-dependent inhibition of the activity of MAO-A and -B respectivey
Catalog No. 50-311-0750 Supplier Selleck Chemical LLC Supplier No. S36901G
Pargyline is an irreversible inhibitor of monoamine oxidase (MAO)-B with Ki values of 13 and 0 5 M for time-dependent inhibition of the activity of MAO-A and -B respectivey