Learn More
Cayman Chemical Pargyline (hydrochloride), 306-07-0, 1 g
SDP
Supplier:  Cayman Chemical 100078521
Encompass_Preferred
An irreversible inhibitor of MAO (Kis = 15 and 1.8 μM for MAO-A and MAO-B, respectively); induces a moderate decrease of systolic blood pressure in unanaesthetized hypertensive rats but not normotensive WKR or Sprague-Dawley rats; prevents reactive oxygen species-mediated monocyte hypertrophy at a concentration of 10 µM,
Catalog No. NC9925372