Learn More
Selleck Chemical LLC Pargyline hydrochloride S3690-25mg
SDP
Catalog No. 501948281
Encompass_Preferred
Pargyline is an irreversible inhibitor of monoamine oxidase (MAO)-B with Ki values of 13 and 0 5 M for time-dependent inhibition of the activity of MAO-A and -B respectivey
Catalog No. 50-194-8281 Supplier Selleck Chemical LLC Supplier No. S369025MG
Pargyline is an irreversible inhibitor of monoamine oxidase (MAO)-B with Ki values of 13 and 0 5 M for time-dependent inhibition of the activity of MAO-A and -B respectivey