Learn More
Medchemexpress LLC Pargyline hydrochloride | 306-07-0 | 99.9% | C11H14ClN | 500 MG
SDP
Catalog No. 5000433482
Encompass_Preferred
Pargyline hydrochloride is a potent and irreversible monoamine oxidase (MAO) inhibitor, demonstrating inhibitory constants (Ki) of 13 μM for MAO-A and 0.5 μM for MAO-B. This compound is recognized for its antihypertensive and anticancer activities.
  • Irreversible MAO inhibition
  • Antihypertensive properties
  • Anticancer activities
  • Click chemistry reagent
  • High purity
  • Versatile solubility
Catalog No. 50-004-33482 Supplier Medchemexpress LLC Supplier No. HYA0091500MG
Pargyline hydrochloride is a potent and irreversible monoamine oxidase (MAO) inhibitor, demonstrating inhibitory constants (Ki) of 13 μM for MAO-A and 0.5 μM for MAO-B. This compound is recognized for its antihypertensive and anticancer activities.
  • Irreversible MAO inhibition
  • Antihypertensive properties
  • Anticancer activities
  • Click chemistry reagent
  • High purity
  • Versatile solubility