Learn More
Selleck Chemical LLC Paricalcitol-1mg
SDP
Catalog No. 502389618
Encompass_Preferred
Paricalcitol (Paracalcin, Zemplar, 19-Nor-1-25-OH2D2, 19-Nor-colecalciferol) is a selective vitamin D receptor agonist. Paricalcitol suppresses parathyroid hormone mRNA expression and inhibits parathyroid cell proliferation. Pariccitol is used to treat secondary hyperparathyroidism in chronic kidney disease.
Catalog No. 50-238-9618 Supplier Selleck Chemical LLC Supplier No. S66811MG
Add to Cart
Add to Cart
Paricalcitol (Paracalcin, Zemplar, 19-Nor-1-25-OH2D2, 19-Nor-colecalciferol) is a selective vitamin D receptor agonist. Paricalcitol suppresses parathyroid hormone mRNA expression and inhibits parathyroid cell proliferation. Pariccitol is used to treat secondary hyperparathyroidism in chronic kidney disease.