Learn More
Medchemexpress LLC Paricalcitol-d6 | 2070009-67-3 | 99.85% | C27H38D6O3 | 1 MG
SDP
Catalog No. 5000434060
Encompass_Preferred
Paricalcitol-d6 is the deuterium labeled Paricalcitol. Paricalcitol is an agent used for the prevention and treatment of secondary hyperparathyroidism (excessive secretion of parathyroid hormone) associated with chronic renal failure. It is for research use only and not sold to patients.
  • This compound can be used as a tracer
  • It can also be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-34060 Supplier Medchemexpress LLC Supplier No. HY765851MG
Paricalcitol-d6 is the deuterium labeled Paricalcitol. Paricalcitol is an agent used for the prevention and treatment of secondary hyperparathyroidism (excessive secretion of parathyroid hormone) associated with chronic renal failure. It is for research use only and not sold to patients.
  • This compound can be used as a tracer
  • It can also be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS