Learn More
Medchemexpress LLC Paris saponin VII | 68124-04-9 | 100.0% | 1031.18 g/mol | C51H82O21 | 10 MG
SDP
Catalog No. 5000289395
Encompass_Preferred
Paris saponin VII is a steroidal saponin (CAS 68124-04-9) isolated from the roots and rhizomes of Trillium species. It is supplied as a high-purity analytical standard for biochemical and cellular research, with reported activity against Akt, p38 MAPK, P-glycoprotein, and Bcl-2 family pathways.
  • High purity analytical standard (99.96%).
  • Molecular formula C51H82O21 and molecular weight 1031.18 g/mol.
  • Reported activity against Akt, p38 MAPK, P-glycoprotein, and Bcl-2 family.
  • Suitable for biochemical assays and cellular studies involving apoptosis and drug resistance.
  • Provided in a 10 mg analytical quantity for research use.
Catalog No. 50-002-89395 Supplier Medchemexpress LLC Supplier No. HYN358410MG
Paris saponin VII is a steroidal saponin (CAS 68124-04-9) isolated from the roots and rhizomes of Trillium species. It is supplied as a high-purity analytical standard for biochemical and cellular research, with reported activity against Akt, p38 MAPK, P-glycoprotein, and Bcl-2 family pathways.
  • High purity analytical standard (99.96%).
  • Molecular formula C51H82O21 and molecular weight 1031.18 g/mol.
  • Reported activity against Akt, p38 MAPK, P-glycoprotein, and Bcl-2 family.
  • Suitable for biochemical assays and cellular studies involving apoptosis and drug resistance.
  • Provided in a 10 mg analytical quantity for research use.