Learn More
Medchemexpress LLC Parishin | 62499-28-9 | 996.91 | 5 MG
SDP
Catalog No. 5000416362
Encompass_Preferred
Parishin is a phenolic glucoside isolated from Gastrodia elata. It exhibits antiaging effects and extends the lifespan of yeast by regulating the Sir2/Uth1/TOR signaling pathway.
  • Exhibits antiaging effects.
  • Extends the lifespan of yeast.
  • Regulates the Sir2/Uth1/TOR signaling pathway.
  • In vitro, extends the replicative lifespan of K6001 yeast at concentrations of 3, 10, and 30 μM.
  • Target: Sir2/Uth1/TOR.
Catalog No. 50-004-16362 Supplier Medchemexpress LLC Supplier No. HYN20315MG
Parishin is a phenolic glucoside isolated from Gastrodia elata. It exhibits antiaging effects and extends the lifespan of yeast by regulating the Sir2/Uth1/TOR signaling pathway.
  • Exhibits antiaging effects.
  • Extends the lifespan of yeast.
  • Regulates the Sir2/Uth1/TOR signaling pathway.
  • In vitro, extends the replicative lifespan of K6001 yeast at concentrations of 3, 10, and 30 μM.
  • Target: Sir2/Uth1/TOR.