Learn More
Medchemexpress LLC Parishin | 62499-28-9 | MFCD22125006 | 98.9% | 996.91 | C45H56O25 | 10 MG
SDP
Catalog No. 5000295937
Encompass_Preferred
Parishin is a phenolic glucoside isolated from Gastrodia elata that exhibits anti-aging activity in yeast via modulation of the Sir2/Uth1/TOR signaling pathway. It is supplied as a high-purity solid with recommended solvent formulations and storage conditions for research applications.
  • High purity (98.92%)
  • Off-white to light yellow solid
  • Available as solid and as 10 mM DMSO solution for in vitro use
  • Recommended storage: 4°C (solid); in solvent: -80°C (6 months), -20°C (1 month)
  • Intended for research use only
Catalog No. 50-002-95937 Supplier Medchemexpress LLC Supplier No. HYN203110MG
Parishin is a phenolic glucoside isolated from Gastrodia elata that exhibits anti-aging activity in yeast via modulation of the Sir2/Uth1/TOR signaling pathway. It is supplied as a high-purity solid with recommended solvent formulations and storage conditions for research applications.
  • High purity (98.92%)
  • Off-white to light yellow solid
  • Available as solid and as 10 mM DMSO solution for in vitro use
  • Recommended storage: 4°C (solid); in solvent: -80°C (6 months), -20°C (1 month)
  • Intended for research use only