Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Parishin B | 174972-79-3 | 20 MG
SDP

Catalog No. 5000421347
Encompass_Preferred

Parishin B is a parishin metabolite isolated from Gastrodia elata with antioxidant activity. It is intended for research use only. This product appears as a white to off-white solid with a purity of 99.42%.

  • Isolated from Gastrodia elata
  • Exhibits antioxidant activity
  • For research use only
  • White to off-white solid appearance
  • Molecular weight of 728.65
  • Chemical formula of C32H40O19
  • Purity of 99.42%
  • Store at 4°C, protected from light
  • For solvent solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Soluble in DMSO and H2O (50 mg/mL)

Catalog No. 50-004-21347 Supplier Medchemexpress LLC Supplier No. HYN212420MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Parishin B is a parishin metabolite isolated from Gastrodia elata with antioxidant activity. It is intended for research use only. This product appears as a white to off-white solid with a purity of 99.42%.

  • Isolated from Gastrodia elata
  • Exhibits antioxidant activity
  • For research use only
  • White to off-white solid appearance
  • Molecular weight of 728.65
  • Chemical formula of C32H40O19
  • Purity of 99.42%
  • Store at 4°C, protected from light
  • For solvent solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Soluble in DMSO and H2O (50 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.