Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Parishin C | 174972-80-6 | 5 MG
SDP

Catalog No. 5000421664
Encompass_Preferred

Parishin C is a parishin derivative isolated from *Gastrodia elata* that may possess antioxidant properties. It is the analytical standard of Parishin C, intended for research and analytical applications.

  • Derived from *Gastrodia elata*
  • May possess antioxidant properties
  • Analytical standard for research and analytical applications
  • Purity: 99.24%
  • Molecular weight: 728.65
  • Molecular formula: C₃₂H₄₀O₁₉
  • Appearance: Solid
  • Color: White to off-white
  • Soluble in DMSO and H₂O
  • Store solid at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-004-21664 Supplier Medchemexpress LLC Supplier No. HYN21255MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Parishin C is a parishin derivative isolated from *Gastrodia elata* that may possess antioxidant properties. It is the analytical standard of Parishin C, intended for research and analytical applications.

  • Derived from *Gastrodia elata*
  • May possess antioxidant properties
  • Analytical standard for research and analytical applications
  • Purity: 99.24%
  • Molecular weight: 728.65
  • Molecular formula: C₃₂H₄₀O₁₉
  • Appearance: Solid
  • Color: White to off-white
  • Soluble in DMSO and H₂O
  • Store solid at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.