Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Parishin E | 952068-57-4 | 1 MG
SDP

Catalog No. 5000421949
Encompass_Preferred

Parishin E is a derivative isolated from Gastrodia elata, identified by its molecular formula C19H24O13 and molecular weight of 460.39. This compound may possess antioxidant properties and is supplied as a white to off-white solid. It is intended for research use only.

  • Isolated from Gastrodia elata
  • Molecular formula: C19H24O13
  • Molecular weight: 460.39
  • Purity: 98.90%
  • Appearance: Solid, white to off-white
  • Classified as a phenol
  • May have antioxidant properties
  • Soluble in DMSO at 100 mg/mL
  • Recommended storage at 4°C, sealed and protected from moisture and light

Catalog No. 50-004-21949 Supplier Medchemexpress LLC Supplier No. HYN21261MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Parishin E is a derivative isolated from Gastrodia elata, identified by its molecular formula C19H24O13 and molecular weight of 460.39. This compound may possess antioxidant properties and is supplied as a white to off-white solid. It is intended for research use only.

  • Isolated from Gastrodia elata
  • Molecular formula: C19H24O13
  • Molecular weight: 460.39
  • Purity: 98.90%
  • Appearance: Solid, white to off-white
  • Classified as a phenol
  • May have antioxidant properties
  • Soluble in DMSO at 100 mg/mL
  • Recommended storage at 4°C, sealed and protected from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.