Learn More
Medchemexpress LLC HY-12594 100mg , Paritaprevir ABT-450;Veruprevir CAS:1216941-48-8 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY12594100MG
Encompass_Preferred
Medchemexpress, HY-12594 100mg Paritaprevir ABT-450;Veruprevir CAS:1216941-48-8 Formula:C40H43N7O7S EC50: 1 nM(HCV 1a), 0.21 nM(HCV 1b) Purity:98% Paritaprevir is a potent non-structural protein 3/4A ( NS3/4A ) protease inhibitor with EC 50 values of 1 and 0.21 nM for HCV 1a and 1b, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2866