Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Parker™ ChromGas™ Zero Air Generator

Catalog No. p-7194241 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Model:
1000
1001
3500
Description:
110 Volt, Output Flow: 1,000 cc/min
110 Volt, Output Flow: 3,500 cc/min
230 Volt, Output Flow: 1,000 cc/min
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Description
11-100-2881 1000 110 Volt, Output Flow: 1,000 cc/min
11-101-6109 1001 230 Volt, Output Flow: 1,000 cc/min
11-100-2763 3500 110 Volt, Output Flow: 3,500 cc/min
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-100-2881 Supplier MilliporeSigma™ Supelco™ Supplier No. 22824
Only null left
Add to Cart
Add to Cart

Eliminates need for expensive gas cylinders

This ChromGas zero air generator incorporates three stages:

  • A 0.5 mm coalescing inlet filter removes particles, oil, and water
  • A heated catalyst removes hydrocarbons
  • A 0.01 mm cellulose fiber outlet filter removes residual particulate material from the product air stream
  • A hazard-free and economical alternative to high pressure gas cylinders, this unit should pay for itself in a year or two.

Features/Benefits

  • Eliminates need for expensive gas cylinders
  • Compact; requires less than one square foot of bench space
  • Can be used anywhere that an electrical supply is available
  • Only requires an upstream compressed air source

Specifications

  • Inlet Pressure: 2-125 psig
  • Outlet Purity: <0.1 ppm hydrocarbons (as methane)
  • Outlet Pressure: 125 psig
  • Outlet Fitting: 1/8 in. compression
  • Power Needs: 110 V (60 Hz), or 230 V (50 Hz)
  • Shipping Weight (Model 1000 and 1001): 11 lb (5 kg)
  • Shipping Weight (Model 3500 and 3501): 20 lb (9.1 kg)
  • Dimensions (Model 1000 and 1001; H x W x D): 9.75 in. x 5.75 in. x 12 in. (25 cm x 14.7 cm x 30.8 cm)
  • Dimensions (Model 3500 and 3501; H x W x D): 12 in. x 6.75 in. x 15 in. (29.2 cm x 17.8 cm x 39.4 cm)

Marks

  • CE approved
  • UL listed
  • CSA listed
  • IEC 1010 certified
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Gas Chromatography
Model 1000
Description 110 Volt, Output Flow: 1,000 cc/min

Parker is a registered trademark of Parker Hannifin

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.