Learn More
MSC Parker Coil Cleaner, 1 gal, Air Cooled Condenser Coils, Bottle
Catalog No. 5000507478
Encompass
This coil cleaner is designed for professional use on air cooled condenser coils. It is supplied in a 1 gallon bottle and is a pink liquid with a pungent odor. This product is corrosive and contains hydrofluoric acid, making it a powerful solution for maintaining refrigeration systems.
  • Professional grade refrigeration coil cleaner.
  • Formulated for air cooled condenser coils.
  • Supplied in a convenient 1 gal bottle.
  • Effectively cleans and maintains condenser coils.
  • Corrosive liquid with a pungent odor.
  • Contains hydrofluoric acid.
Catalog No. 50-005-07478 Supplier MSC Supplier No. 80216237
This coil cleaner is designed for professional use on air cooled condenser coils. It is supplied in a 1 gallon bottle and is a pink liquid with a pungent odor. This product is corrosive and contains hydrofluoric acid, making it a powerful solution for maintaining refrigeration systems.
  • Professional grade refrigeration coil cleaner.
  • Formulated for air cooled condenser coils.
  • Supplied in a convenient 1 gal bottle.
  • Effectively cleans and maintains condenser coils.
  • Corrosive liquid with a pungent odor.
  • Contains hydrofluoric acid.