Learn More
MSC Parker Coil Cleaner, Alkaline, 1 gal
Catalog No. 5000507568
Encompass
This coil cleaner is an alkaline formula designed to effectively remove bacteria, bugs, lint, and scale from evaporator coils and drain pans. It is biodegradable, making it a suitable choice for various applications.
  • Alkaline formula
  • Removes bugs, lint, bacteria, and scale
  • Suitable for evaporator coils and drain pans
  • Biodegradable
Catalog No. 50-005-07568 Supplier MSC Supplier No. 80216278
This coil cleaner is an alkaline formula designed to effectively remove bacteria, bugs, lint, and scale from evaporator coils and drain pans. It is biodegradable, making it a suitable choice for various applications.
  • Alkaline formula
  • Removes bugs, lint, bacteria, and scale
  • Suitable for evaporator coils and drain pans
  • Biodegradable