Learn More
MSC Parker Hannifin Elastic Sealant Compound, Non-Hardening, Water-Resistant, 2 lb
Catalog No. 5000507479
Encompass
This elastic sealant compound is designed to provide an effective barrier against water, moisture, dirt, and dust in various applications. It is non-hardening, water-resistant, and formulated from high-quality materials. This sealant offers excellent adhesion to clean, dry surfaces and remains permanently pliable.
  • Non-hardening
  • Water-resistant
  • Formulated from the highest quality materials
  • Non-staining
  • Non-bleeding
  • Excellent adhesion to clean dry surfaces
  • Supplied in ready-to-use extruded cords or bulk compound
  • Permanently pliable
  • Provides an effective barrier against water, moisture, dirt and dust
Catalog No. 50-005-07479 Supplier MSC Supplier No. 80217250
This elastic sealant compound is designed to provide an effective barrier against water, moisture, dirt, and dust in various applications. It is non-hardening, water-resistant, and formulated from high-quality materials. This sealant offers excellent adhesion to clean, dry surfaces and remains permanently pliable.
  • Non-hardening
  • Water-resistant
  • Formulated from the highest quality materials
  • Non-staining
  • Non-bleeding
  • Excellent adhesion to clean dry surfaces
  • Supplied in ready-to-use extruded cords or bulk compound
  • Permanently pliable
  • Provides an effective barrier against water, moisture, dirt and dust