Learn More
MSC Parker Ice Machine Cleaner, Metal Safe, 16 oz, Nickel/Tin Evaporator Safe
Catalog No. 5000507620
Encompass
This ice machine cleaner and scale remover is designed for maintaining good ice quality and efficient cooling tower systems, which is crucial for various commercial establishments. It provides effective cleaning and scale removal to accomplish this task quickly and easily.
  • Bottle container type.
  • 16 oz capacity.
  • Metal-safe cleaner formulation.
  • Safe for use on nickel/tin plated evaporators.
  • Suitable for ice machines (cube, tube, flake types) and commercial dishwashers.
  • Functions as both an ice machine cleaner and scale remover.
  • Contributes to energy efficiency.
  • Helps increase ice production.
  • Contains phosphoric acid and glycolic acid as active ingredients.
Catalog No. 50-005-07620 Supplier MSC Supplier No. 80217102
This ice machine cleaner and scale remover is designed for maintaining good ice quality and efficient cooling tower systems, which is crucial for various commercial establishments. It provides effective cleaning and scale removal to accomplish this task quickly and easily.
  • Bottle container type.
  • 16 oz capacity.
  • Metal-safe cleaner formulation.
  • Safe for use on nickel/tin plated evaporators.
  • Suitable for ice machines (cube, tube, flake types) and commercial dishwashers.
  • Functions as both an ice machine cleaner and scale remover.
  • Contributes to energy efficiency.
  • Helps increase ice production.
  • Contains phosphoric acid and glycolic acid as active ingredients.