Learn More
MSC Parker Ice Machine Cleaner, Phosphoric Acid, Professional Use
Catalog No. 5000508889
Encompass
This ice machine cleaner from Parker is an aqueous acid solution specifically designed for professional use in cleaning ice machines. It features a high concentration of phosphoric acid, making it effective for tackling scale and buildup, ensuring optimal performance and hygiene of the equipment.
  • Designed for professional ice machine cleaning
  • Formulated as an aqueous acid solution
  • Contains phosphoric acid, 70-80% concentration
  • Effective for scale removal
  • Odorless clear liquid
  • Low pH of 1-2
Catalog No. 50-005-08889 Supplier MSC Supplier No. 60513942
This ice machine cleaner from Parker is an aqueous acid solution specifically designed for professional use in cleaning ice machines. It features a high concentration of phosphoric acid, making it effective for tackling scale and buildup, ensuring optimal performance and hygiene of the equipment.
  • Designed for professional ice machine cleaning
  • Formulated as an aqueous acid solution
  • Contains phosphoric acid, 70-80% concentration
  • Effective for scale removal
  • Odorless clear liquid
  • Low pH of 1-2