Learn More
MSC Parker Liquid Scale Remover, 1 Gallon, Heavily Scaled Equipment, with Inhibitors
Catalog No. 5000507462
Encompass
This liquid scale remover is formulated for cleaning heavily scaled equipment and effectively removing various problem scales. It incorporates anti-foaming agents and corrosion inhibitors to protect metal surfaces during the cleaning process. This product is intended for professional use, especially as a refrigeration coil cleaner.
  • Formulated for heavily scaled equipment and problem scales.
  • Contains anti-foaming agents for efficient cleaning.
  • Includes inhibitors to minimize metal damage.
  • Maintains good ice quality and efficient cooling tower systems.
  • Provides effective cleaning and scale removal.
  • Designed for quick and easy job completion.
  • Professional use only, as a refrigeration coil cleaner.
Catalog No. 50-005-07462 Supplier MSC Supplier No. 80217219
This liquid scale remover is formulated for cleaning heavily scaled equipment and effectively removing various problem scales. It incorporates anti-foaming agents and corrosion inhibitors to protect metal surfaces during the cleaning process. This product is intended for professional use, especially as a refrigeration coil cleaner.
  • Formulated for heavily scaled equipment and problem scales.
  • Contains anti-foaming agents for efficient cleaning.
  • Includes inhibitors to minimize metal damage.
  • Maintains good ice quality and efficient cooling tower systems.
  • Provides effective cleaning and scale removal.
  • Designed for quick and easy job completion.
  • Professional use only, as a refrigeration coil cleaner.