Learn More
Parker Hannifan Parker N2-20PBLN-NA-C-LAB Nitrogen Gas Generator, High Purity Nitrogen Production, Energy Efficient PSA Technology
SDP
Catalog No. NC3300496
Encompass
The Parker N2-20PBLN-NA-C-LAB is a highly efficient nitrogen gas generator using Pressure Swing Adsorption (PSA) technology for on-site production of high-purity nitrogen from compressed air. It offers significant cost savings and reliability compared to traditional nitrogen supply methods. With adjustable purity levels up to 99.9995%, it supports various industrial applications, ensuring compliance with food and pharmaceutical standards, and features a robust, modular design for scalability.
Catalog No. NC3300496 Supplier Parker Hannifan Supplier No. N220PBLNNACLAB
Add to Cart
Add to Cart
Request a Quote
The Parker N2-20PBLN-NA-C-LAB is a highly efficient nitrogen gas generator using Pressure Swing Adsorption (PSA) technology for on-site production of high-purity nitrogen from compressed air. It offers significant cost savings and reliability compared to traditional nitrogen supply methods. With adjustable purity levels up to 99.9995%, it supports various industrial applications, ensuring compliance with food and pharmaceutical standards, and features a robust, modular design for scalability.