Learn More
Bioworld Parker Thompson Fern Basal Salt Mixture, 50 L
Catalog No. 50254836
Encompass_Preferred
Parker Thompson Fern Basal Salt MixtureContains the Macronutrients and Micronutrients as described by Hickok et all. (1995) Formulated for the growth of Ceratopteris RichardiiSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of mediumPlant tissue culture testedWorking Concentration: 0.77 g/LpH: 3.5–4.5Soluble in water
Catalog No. 50-254-836 Supplier Bioworld Supplier No. 306301393
Parker Thompson Fern Basal Salt MixtureContains the Macronutrients and Micronutrients as described by Hickok et all. (1995) Formulated for the growth of Ceratopteris RichardiiSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of mediumPlant tissue culture testedWorking Concentration: 0.77 g/LpH: 3.5–4.5Soluble in water