Learn More
Perkin Elmer US LLC Parker-Hannifin Nut and Ferrule Kit - 0.0625 in, 6 ea.
Catalog No. 502112091
Encompass_Preferred
Parker-Hannifin stainless steel nuts are used to connect 1/16 in O.D. stainless steel tubing and feature a 10 32 thread size. This kit includes the following pieces: 6 Stainless Steel Parker-Hannifin Ferrules, 1/16 inch I.D. (09903974) 6 Inverted Tubing Nuts, 1/16 inch (09903980)
Catalog No. 50-211-2091 Supplier Perkin Elmer US LLC Supplier No. 00890945
Parker-Hannifin stainless steel nuts are used to connect 1/16 in O.D. stainless steel tubing and feature a 10 32 thread size. This kit includes the following pieces: 6 Stainless Steel Parker-Hannifin Ferrules, 1/16 inch I.D. (09903974) 6 Inverted Tubing Nuts, 1/16 inch (09903980)