Learn More
Enzo Life Sciences Parkin (human) polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW93650100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 399-415 (CE339QARWEAASKETIK412) of human parkin. A single amino acid substitution (A405E) exists in both the rat and mouse homologues. Species reactivity: Human. Applications: IHC, IP, WB. Formulation: Liquid. In PBS.
Catalog No. 50-201-1682