Learn More
Mettler Toledo™ Parking Beaker for EasyPlace Electrode Arm
Catalog No. 01911933 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
EasyPlace
Catalog No. For Use With (Equipment)
01-911-933 EasyPlace
1 options
This item is not returnable. View return policy
Catalog No. 01-911-933 Supplier Mettler Toledo™ Supplier No. 30671571
Add to Cart
Add to Cart
This item is not returnable. View return policy
Parking beaker for the EasyPlace electrode arm. Fill it with storage solution and keep your electrode perfectly conditioned also in short breaks between measurements, or use it to store the wetting cap while measuring.
  • Safe Short-Term Storage
  • The parking beaker (with insert) stores a sensor in a suitable storage solution without removing it from the EasyPlace - never store your sensor dry
TRUSTED_SUSTAINABILITY
Specifications
Type Parking Beaker, EasyPlace
For Use With (Equipment) EasyPlace