Learn More
Mettler Toledo™ Parking Beaker Insert, EasyPlace
Catalog No. 01911934 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
EasyPlace
Catalog No. For Use With (Equipment)
01-911-934 EasyPlace
1 options
This item is not returnable. View return policy
Catalog No. 01-911-934 Supplier Mettler Toledo™ Supplier No. 30671572
Add to Cart
Add to Cart
This item is not returnable. View return policy
The parking beaker insert should be used while using the parking beaker with storage solution.
  • Safe Short-Term Storage
  • The parking beaker insert holds storage solutions and can be placed in the parking beaker
TRUSTED_SUSTAINABILITY
Specifications
Type Parking Beaker Insert, EasyPlace
For Use With (Equipment) EasyPlace