Learn More
Aviva Systems Biology Corp PARL Antibody - N-terminal region (ARP44851_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44851_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PARL. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PARL. Alternate names: PRO2207, PSARL, PSARL1, PSENIP2, RHBDS1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53437