Learn More
Electron Microscopy Sciences Parlodion (Pyroxylin Purified Strips) 10 GR
SDP
Catalog No. 5027695
Encompass_Preferred
CAS #9004-70-0 C₁₂H₁₆N₄O₁₈ F.W. 504.28
Highly purified from cellulose nitrate; specially prepared for embedding tissue for sectioning and for preparing semi-permeable membranes. In strips.
Catalog No. 50-276-95 Supplier Electron Microscopy Sciences Supplier No. 19220
CAS #9004-70-0 C₁₂H₁₆N₄O₁₈ F.W. 504.28
Highly purified from cellulose nitrate; specially prepared for embedding tissue for sectioning and for preparing semi-permeable membranes. In strips.