Learn More
Medchemexpress LLC HY-13965 5mg Medchemexpress, Parmodulin 2 CAS:423735-93-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY139655MG
Encompass_Preferred
Medchemexpress, HY-13965 5mg Parmodulin 2 CAS:423735-93-7 Parmodulin 2 (ML161),a probe, is an allosteric inhibitor of protease-activated receptor 1 (PAR1) with an IC50 of 0.26 μM. Parmodulin 2 inhibits platelet aggregation induced by a PAR1 peptide agonist or by thrombin and has shown cytoprotective effects[1, 2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9891