Learn More
Aviva Systems Biology Corp PARN Antibody - N-terminal region (ARP37393_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37393_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PARN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: DAN, 1200003I18RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56361