Learn More
Alkali Scientific Paromomycin Sulfate, 1 G
SDP
Catalog No. 5000659999
Encompass_Preferred
Paromomycin Sulfate is an aminoglycoside antibiotic used in a variety of microbiological applications. This 1 g powdered formulation is effective in treating infections caused by protozoa and certain bacteria. It works by binding to the bacterial ribosome, inhibiting protein synthesis, and ultimately leading to cell death. Paromomycin is often used in laboratory studies to evaluate the effectiveness of antibiotics against resistant strains and to investigate the role of antibiotics in antimicrobial resistance. This 1 g formulation is widely used in pharmaceutical testing and clinical microbiology to treat infections caused by susceptible organisms and to test for resistance patterns in microbial cultures.
Catalog No. 50-006-59999 Supplier Alkali Scientific Supplier No. PAR002CP
Paromomycin Sulfate is an aminoglycoside antibiotic used in a variety of microbiological applications. This 1 g powdered formulation is effective in treating infections caused by protozoa and certain bacteria. It works by binding to the bacterial ribosome, inhibiting protein synthesis, and ultimately leading to cell death. Paromomycin is often used in laboratory studies to evaluate the effectiveness of antibiotics against resistant strains and to investigate the role of antibiotics in antimicrobial resistance. This 1 g formulation is widely used in pharmaceutical testing and clinical microbiology to treat infections caused by susceptible organisms and to test for resistance patterns in microbial cultures.