Learn More
Medchemexpress LLC Paromomycin sulfate | 1263-89-4 | 100.0% | 500 MG
SDP
Catalog No. 5000408182
Encompass_Preferred
Paromomycin sulfate (Aminosidine sulfate) is a broad-spectrum aminoglycoside antibiotic and a derivative of neomycin. It functions by prematurely terminating mRNA translation and inhibiting protein synthesis through specific binding to the RNA oligonucleotide at the A site of bacterial 30S ribosomes. This compound demonstrates both amebicidal and bactericidal properties, making it valuable for research into bacterial and parasitic infections.
  • Broad-spectrum aminoglycoside antibiotic activity
  • Exhibits amebicidal and bactericidal properties
  • Inhibits protein synthesis by targeting bacterial 30S ribosomes
  • Useful for research on bacterial and parasitic infections
  • Reduces intracellular parasitic forms in Caco-2 and HCT-8 cells in vitro
  • Reduces C. parvum oocysts in infected mice in vivo
Catalog No. 50-004-08182 Supplier Medchemexpress LLC Supplier No. HYB0956500MG
Paromomycin sulfate (Aminosidine sulfate) is a broad-spectrum aminoglycoside antibiotic and a derivative of neomycin. It functions by prematurely terminating mRNA translation and inhibiting protein synthesis through specific binding to the RNA oligonucleotide at the A site of bacterial 30S ribosomes. This compound demonstrates both amebicidal and bactericidal properties, making it valuable for research into bacterial and parasitic infections.
  • Broad-spectrum aminoglycoside antibiotic activity
  • Exhibits amebicidal and bactericidal properties
  • Inhibits protein synthesis by targeting bacterial 30S ribosomes
  • Useful for research on bacterial and parasitic infections
  • Reduces intracellular parasitic forms in Caco-2 and HCT-8 cells in vitro
  • Reduces C. parvum oocysts in infected mice in vivo