Learn More
Apexbio Technology LLC Paromomycin Sulfate 1263-89-4 10mM (in 1mL H2O)
SDP
Catalog No. 5000094667
Encompass_Preferred
Paromomycin Sulfate (CAS 1263-89-4) is a small-molecule antibiotic targeting the 16S rRNA region of the bacterial 30S ribosomal subunit It is designed to disrupt messenger RNA decoding thereby inhibiting protein biosynthesis in susceptible cells Paromomycin Sulfate exerts its biological activity primarily through binding to the bacterial 30S ribosomal subunit In in vitro studies Paromomycin Sulfate demonstrates inhibition with reported IC50 values typically ranging between 5 and 15 M varying depending on the organism and experimental conditions Based on these pharmacological properties Paromomycin Sulfate holds research potential in the selection of genetically engineered organisms investigation of ribosomal function and the study of antibiotic resistance mechanisms
Catalog No. 50-000-94667 Supplier Apexbio Technology LLC Supplier No. B18085.1
Paromomycin Sulfate (CAS 1263-89-4) is a small-molecule antibiotic targeting the 16S rRNA region of the bacterial 30S ribosomal subunit It is designed to disrupt messenger RNA decoding thereby inhibiting protein biosynthesis in susceptible cells Paromomycin Sulfate exerts its biological activity primarily through binding to the bacterial 30S ribosomal subunit In in vitro studies Paromomycin Sulfate demonstrates inhibition with reported IC50 values typically ranging between 5 and 15 M varying depending on the organism and experimental conditions Based on these pharmacological properties Paromomycin Sulfate holds research potential in the selection of genetically engineered organisms investigation of ribosomal function and the study of antibiotic resistance mechanisms