Learn More
Medchemexpress LLC Paromomycin sulfate | 1263-89-4 | 99.9% | 1 ML
SDP
Catalog No. 5000408956
Encompass_Preferred
Paromomycin sulfate is a neomycin derivative, functioning as a broad-spectrum aminoglycoside antibiotic with both amebicidal and bactericidal effects. It primarily works by prematurely terminating mRNA translation and inhibiting protein synthesis through specific binding to the RNA oligonucleotide at the A site of bacterial 30S ribosomes. This compound is utilized in the research of various bacterial and parasitic infections.
  • Broad-spectrum aminoglycoside antibiotic
  • Exhibits amebicidal and bactericidal effects
  • Inhibits protein synthesis by targeting bacterial 30S ribosomes
  • Useful in research for bacterial and parasitic infections
  • Demonstrates reduction in intracellular parasitic forms in cell cultures
  • Shown to reduce oocysts in vivo models
Catalog No. 50-004-08956 Supplier Medchemexpress LLC Supplier No. HYB095610MM/1ML
Paromomycin sulfate is a neomycin derivative, functioning as a broad-spectrum aminoglycoside antibiotic with both amebicidal and bactericidal effects. It primarily works by prematurely terminating mRNA translation and inhibiting protein synthesis through specific binding to the RNA oligonucleotide at the A site of bacterial 30S ribosomes. This compound is utilized in the research of various bacterial and parasitic infections.
  • Broad-spectrum aminoglycoside antibiotic
  • Exhibits amebicidal and bactericidal effects
  • Inhibits protein synthesis by targeting bacterial 30S ribosomes
  • Useful in research for bacterial and parasitic infections
  • Demonstrates reduction in intracellular parasitic forms in cell cultures
  • Shown to reduce oocysts in vivo models