Learn More
Medchemexpress LLC Paromomycin sulfate | 1263-89-4 | MFCD00079278 | 99.9% | 5 G
SDP
Catalog No. 5000409135
Encompass_Preferred
Paromomycin sulfate is a broad-spectrum aminoglycoside antibiotic, a neomycin derivative, known for its amebicidal and bactericidal effects. It functions by prematurely terminating mRNA translation and inhibiting protein synthesis by specifically binding to the RNA oligonucleotide at the A site of bacterial 30S ribosomes.
  • Can be used for the research of bacterial infections
  • Can be used for the research of parasitic infections
  • Effective for acute and chronic intestinal protozoal infections
  • Used as a therapeutic against visceral leishmaniasis
  • Exhibits antibacterial, antiprotozoal, anthelminthic, and antiparasitic effects
Catalog No. 50-004-09135 Supplier Medchemexpress LLC Supplier No. HYB09565G
Paromomycin sulfate is a broad-spectrum aminoglycoside antibiotic, a neomycin derivative, known for its amebicidal and bactericidal effects. It functions by prematurely terminating mRNA translation and inhibiting protein synthesis by specifically binding to the RNA oligonucleotide at the A site of bacterial 30S ribosomes.
  • Can be used for the research of bacterial infections
  • Can be used for the research of parasitic infections
  • Effective for acute and chronic intestinal protozoal infections
  • Used as a therapeutic against visceral leishmaniasis
  • Exhibits antibacterial, antiprotozoal, anthelminthic, and antiparasitic effects