Learn More
Alkali Scientific Paromomycin Sulfate, 25 G
SDP
Catalog No. 5000660998
Encompass_Preferred
Paromomycin Sulfate, 25 g, is an antibiotic used to treat infections caused by protozoa and certain bacteria. This formulation is commonly used in laboratory studies to evaluate antimicrobial activity and resistance. Paromomycin is an aminoglycoside antibiotic that targets bacterial protein synthesis, making it effective against a broad spectrum of organisms. In research, it is used for susceptibility testing, drug development, and antimicrobial resistance studies. The 25 g powder format is ideal for preparing larger volumes of working solution for in vitro testing or clinical applications, particularly in pharmaceutical and microbiological research environments where more substantial quantities of antibiotic are needed.
Catalog No. 50-006-60998 Supplier Alkali Scientific Supplier No. PAR003CP
Paromomycin Sulfate, 25 g, is an antibiotic used to treat infections caused by protozoa and certain bacteria. This formulation is commonly used in laboratory studies to evaluate antimicrobial activity and resistance. Paromomycin is an aminoglycoside antibiotic that targets bacterial protein synthesis, making it effective against a broad spectrum of organisms. In research, it is used for susceptibility testing, drug development, and antimicrobial resistance studies. The 25 g powder format is ideal for preparing larger volumes of working solution for in vitro testing or clinical applications, particularly in pharmaceutical and microbiological research environments where more substantial quantities of antibiotic are needed.