Learn More
Research Products International Corp PAROMOMYCIN SULFATE, 25 GM
SDP
Catalog No. 50213591
Encompass_Preferred
Paromomycin Sulfate belongs to the aminoglycoside class of antibiotics. It is primarily studied for its antibiotic properties, particularly its effectiveness against various types of bacteria and parasites.
Researchers investigate its mechanism of action, pharmacokinetics, and potential applications in treating infectious diseases.
Paromomycin is often used as a tool in molecular biology research due to its ability to inhibit protein synthesis in bacteria and parasites. It is employed in experiments involving gene expression, translation studies, and the development of antibiotic resistance mechanisms.
Catalog No. 50-213-591 Supplier Research Products International Corp Supplier No. P1100025.0
Paromomycin Sulfate belongs to the aminoglycoside class of antibiotics. It is primarily studied for its antibiotic properties, particularly its effectiveness against various types of bacteria and parasites.
Researchers investigate its mechanism of action, pharmacokinetics, and potential applications in treating infectious diseases.
Paromomycin is often used as a tool in molecular biology research due to its ability to inhibit protein synthesis in bacteria and parasites. It is employed in experiments involving gene expression, translation studies, and the development of antibiotic resistance mechanisms.