Learn More
Selleck Chemical LLC Paromomycin Sulfate S4183-50mg
SDP
Catalog No. 503109124
Encompass_Preferred
Paromomycin Sulfate(Aminosidine sulfate) is an aminoglycoside antibiotics inhibiting protein synthesis in non-resistant cells by binding to 16S ribosomal RNA
Catalog No. 50-310-9124 Supplier Selleck Chemical LLC Supplier No. S418350MG
Paromomycin Sulfate(Aminosidine sulfate) is an aminoglycoside antibiotics inhibiting protein synthesis in non-resistant cells by binding to 16S ribosomal RNA