Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paromomycin sulfate, 98.3%, MP Biomedicals™

Catalog No. ICN15605625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15605625 25 g
ICN15605601 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15605625 Supplier MP Biomedicals, Inc Supplier No. 0215605625
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1263-89-4
Molecular Formula C23H47N5O18S
Molecular Weight (g/mol) 713.71
InChI Key LJRDOKAZOAKLDU-UMIPPVEZSA-N
Synonym Paromomycin sulfate salt
PubChem CID 4696
ChEBI CHEBI:74926
SMILES OS(O)(=O)=O.NC[C@@H]1O[C@H](O[C@@H]2[C@@H](CO)OC(O[C@@H]3[C@@H](O)[C@H](N)C[C@H](N)C3O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)[C@H](N)[C@@H](O)[C@@H]1O

Specifications

Type Paromomycin Sulfate
Physical Form Powder
Color White
Quantity 25 g
Assay Percent Range >97%
IUPAC Name 4-hydroxy-4,6-dihydrofuro[3,2-c]pyran-2-one
Formula Weight 615.6
Percent Purity 98.3%
Grade Not Available
Chemical Name or Material Paromomycin Sulfate
EINECSNumber 215-031-7
Recommended Storage Store at room temperature (15° to 30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.