Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paromomycin Sulfate, Fisher BioReagents

Catalog No. BP29531 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP29531 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP29531

missing translation for 'validMainframeMsg'

Inhibits protein synthesis

  • A broad spectrum aminoglycoside antibiotic
  • Functions as a protein synthesis inhibitor
  • Useful in the study of bacterial protein synthesis, RNA molecules, and the effects of antibiotics on cell lines
  • Suitable for cell culture applications; research only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1263-89-4
Molecular Formula C23H47N5O18S
Molecular Weight (g/mol) 713.706
InChI Key LJRDOKAZOAKLDU-JXDXLELXSA-N
PubChem CID 91972180
IUPAC Name (3S,5S,6S)-5-amino-2-(aminomethyl)-6-[(2R,4S,5S)-5-[(2R,5R,6R)-3,5-diamino-2-[(2S,3S,5S)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-hydroxycyclohexyl]oxy-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxyoxane-3,4-diol;sulfuric acid
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)O)N)OC3C(C(C(O3)CO)OC4C(C(C(C(O4)CN)O)O)N)O)O)N.OS(=O)(=O)O

Specifications

Type Paromomycin Sulfate
Physical Form Powder/Solid
Source Streptomyces krestomuceticus
Loss on Drying < 5%
Packaging Amber Glass
Potency > 675 U/mg
Quantity 1 g
Formula Weight 615.6
Chemical Name or Material Paromomycin Sulfate
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes, respiratory system and skin. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:2
Flammability:0
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.