Learn More
Paromomycin Sulfate, USP Grade, G-Biosciences
SCI_ED
Catalog No. S139806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139806 25 g
S139737 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139806 Supplier G-Biosciences Supplier No. RC1083
Add to Cart
Add to Cart
Paromomycin is an aminoglycoside antibiotic isolated from Streptomyces rimosus. It targets a variety of gram-negative and gram-positive bacteria as well as several protozoan species.
It displays activity similar to that of neomycin. Paromomycin sulfate is freely soluble.
Specifications
Form Powder
Product Type Antimicrobial
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Quantity 25 g