Learn More
Cayman Chemical Paromomycin (sulfate), Purity: ≥98%, CAS: 1263-89-4, 25 g
SDP
Catalog No. 502964381
Encompass_Preferred
An aminoglycoside antibiotic; effective against Gram-negative and Gram-positive bacteria (MICs = 0.08-3.9 µg/ml); antiprotozoal; decreases the parasite burden of mice infected with L. tropica when administered at a dose of 50 mg/kg and 15 mg/kg, through intramuscular or intralesional routes, respectively; binds to the tRNA decoding A site of the 16S ribosomal RNA in bacteria; inhibits protein synthesis
Catalog No. 50-296-4381 Supplier Cayman Chemical Supplier No. 2363425 G
An aminoglycoside antibiotic; effective against Gram-negative and Gram-positive bacteria (MICs = 0.08-3.9 µg/ml); antiprotozoal; decreases the parasite burden of mice infected with L. tropica when administered at a dose of 50 mg/kg and 15 mg/kg, through intramuscular or intralesional routes, respectively; binds to the tRNA decoding A site of the 16S ribosomal RNA in bacteria; inhibits protein synthesis