Learn More
Medchemexpress LLC Paroxetine-d4 hydrochloride | 2714485-95-5 | 99.2% | 1 MG
SDP
Catalog No. 5000405267
Encompass_Preferred
Paroxetine-d4 hydrochloride is a deuterium-labeled version of Paroxetine hydrochloride, a potent selective serotonin-reuptake inhibitor used as an antidepressant. It has GRK2 inhibitory ability with an IC50 of 14 μM and is suitable for research on depressive disorders. Deuterium-labeled drug molecules are used as tracers for quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05267 Supplier Medchemexpress LLC Supplier No. HYB0492S11MG
Add to Cart
Add to Cart
Paroxetine-d4 hydrochloride is a deuterium-labeled version of Paroxetine hydrochloride, a potent selective serotonin-reuptake inhibitor used as an antidepressant. It has GRK2 inhibitory ability with an IC50 of 14 μM and is suitable for research on depressive disorders. Deuterium-labeled drug molecules are used as tracers for quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS