Learn More
Medchemexpress LLC HY-B0492 100mg Medchemexpress, Paroxetine (hydrochloride) CAS:78246-49-8 Purity:>98%
SDP
Catalog No. 502082563
Encompass_Preferred
Medchemexpress, HY-B0492 100mg Paroxetine (hydrochloride) CAS:78246-49-8 Paroxetine hydrochloride is a potent selective serotonin-reuptake inhibitor, commonly prescribed as an antidepressant and has GRK2 inhibitory ability with IC50 of 14 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2563 Supplier Medchemexpress LLC Supplier No. HYB0492100MG
Medchemexpress, HY-B0492 100mg Paroxetine (hydrochloride) CAS:78246-49-8 Paroxetine hydrochloride is a potent selective serotonin-reuptake inhibitor, commonly prescribed as an antidepressant and has GRK2 inhibitory ability with IC50 of 14 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.