Learn More
TARGETMOL CHEMICALS INC Paroxetine hydrochloride 200MG
SDP
Catalog No. 502877755
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Paroxetine hydrochloride (Paroxetine HCl) is a serotonin uptake inhibitor that is effective in the treatment of depression. Purity 99.91%
Catalog No. 50-287-7755 Supplier TARGETMOL CHEMICALS INC Supplier No. T1636200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Paroxetine hydrochloride (Paroxetine HCl) is a serotonin uptake inhibitor that is effective in the treatment of depression. Purity 99.91%